missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3,4-difluoroaniline, 97%

Catalog No. AAH6187614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6187614 25 g
AAH6187606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6187614 Supplier Thermo Scientific Chemicals Supplier No. H6187614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1092349-87-5 | C6H4BrF2N | 208.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1092349-87-5
Molecular Formula C6H4BrF2N
Molecular Weight (g/mol) 208.01
MDL Number MFCD11035924
InChI Key ZGZURPPCPVPDMX-UHFFFAOYSA-N
Synonym 2-bromo-3,4-difluoro aniline, 2-bromo-3,4-difluorophenylamine, 2-bromo-3,4-difluorobenzenamine
PubChem CID 22731604
IUPAC Name 2-bromo-3,4-difluoroaniline
SMILES NC1=C(Br)C(F)=C(F)C=C1

Specifications

Quantity 25 g
UN Number UN2811
Formula Weight 208
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Bromo-3,4-difluoroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only