missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-(2-ethylhexyl)thiophene 98.0+%, TCI America™

Catalog No. B47831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47831G 1 g
B47835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47831G Supplier TCI America Supplier No. B47831G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 303734-52-3
Molecular Formula C12H19BrS
Molecular Weight (g/mol) 275.248
MDL Number MFCD26743667
InChI Key VWIAAXXJIUXUEC-UHFFFAOYSA-N
PubChem CID 58098291
IUPAC Name 2-bromo-3-(2-ethylhexyl)thiophene
SMILES CCCCC(CC)CC1=C(SC=C1)Br

Specifications

Color Yellow
Quantity 1 g
Formula Weight 275.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-(2-ethylhexyl)thiophene
TSCA No
Recommended Storage Refrigerator