missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-nitro-1,3-propanediol, 98%

Catalog No. AC158820250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158820250 25 g Plastic bottle
AC158821000 100 g Plastic bottle
AC158825000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC158820250 Supplier Thermo Scientific Chemicals Supplier No. 158820250

CAS: 52-51-7 | C3H6BrNO4 | 199.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52-51-7
Molecular Formula C3H6BrNO4
Molecular Weight (g/mol) 199.99
MDL Number MFCD00007390
InChI Key LVDKZNITIUWNER-UHFFFAOYSA-N
Synonym bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol
PubChem CID 2450
ChEBI CHEBI:31306
IUPAC Name 2-bromo-2-nitropropane-1,3-diol
SMILES C(C(CO)([N+](=O)[O-])Br)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >120.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula HOCH2C(Br)(NO2)CH2OH
Quantity 25 g
Beilstein 01, IV, 2501
Merck Index 15, 1457
Solubility Information Solubility in water: 250g/L at 22°C. Other solubilities: soluble in ethanol (500g/L at 24°C),glycerol,(10g/L at 23°C),propylene glycol (400g/L at,23°C),ethyl acetate;,slightly soluble in chcl3,acetone,ether,benzene
Formula Weight 199.99
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Bromo-2-nitro-1, 3-propanediol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 200-143-

RUO – Research Use Only