missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-méthyl-2-phényl-1,3-thiazol-5-yl)-1-éthanone, 95%, Thermo Scientific™

Numéro de catalogue. CC29205DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC29205DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC29205DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC29205DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 7520-95-8
Formule moléculaire C12H10BrNOS
Poids moléculaire (g/mol) 296.182
Numéro MDL MFCD04071443
Clé InChI BOMSILGSXFNLJX-UHFFFAOYSA-N
Synonyme 2-bromo-1-4-methyl-2-phenyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-phenyl-1,3-thiazol-5-yl ethanone, 5-bromoacetyl-4-methyl-2-phenylthiazole, 2-bromo-1-4-methyl-2-phenylthiazol-5-yl ethanone, 2-bromo-1-4-methyl-2-phenyl 1,3-thiazol-5-yl ethan-1-one
PubChem CID 2795492
Nom de l’IUPAC 2-bromo-1-(4-méthyl-2-phényl-1,3-thiazol-5-yl)éthanone
SOURIRES CC1=C(SC(=N1)C2=CC=CC=C2)C(=O)CBr

Spécifications

Nom ou substance chimique 2-Bromo-1-(4-methyl-2-phenyl-1, 3-thiazol-5-yl)-1-ethanone
CAS % minimum 89.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 90%
Emballage Bouteille en verre ambrée
Quantité 1 g
Poids de formule 296.19
Pourcentage de pureté 90%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.