missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethan-1-one, Thermo Scientific™

Catalog No. CC01305DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC01305DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC01305DA Supplier Thermo Scientific Chemicals Supplier No. CC01305DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4629-54-3
Molecular Formula C10H9BrO3
Molecular Weight (g/mol) 257.08
MDL Number MFCD00099402
InChI Key CSSHRKYOZTZFCX-UHFFFAOYSA-N
Synonym 2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl ethanone, 2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl ethan-1-one, 2-bromo-1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethanone, 2-bromo-1-2,3-dihydrobenzo 1,4 dioxin-6-yl ethanone, 2-bromo-1-2,3-dihydrobenzo 1,4-dioxin-6-yl ethanone, 1,4-benzodioxan-6-ylbromomethyl ketone, 2-bromo-1-2,3-dihydro-benzo 1,4 dioxin-6-yl-ethanone, ethanone,2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl
PubChem CID 2776171
SMILES BrCC(=O)C1=CC=C2OCCOC2=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 2-bromo-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethanone
Formula Weight 257.08
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(2, 3-dihydro-1, 4-benzodioxin-6-yl)ethan-1-one