missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(1,3-thiazol-2-yl)ethanone, ≥97%, Thermo Scientific™

Catalog No. CC09505DA
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC09505DA 1 g
CC09505CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC09505DA Supplier Thermo Scientific Chemicals Supplier No. CC09505DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3292-77-1
Molecular Formula C5H4BrNOS
Molecular Weight (g/mol) 206.06
MDL Number MFCD06411540
InChI Key AQRFTRDAOYSMEA-UHFFFAOYSA-N
Synonym 2-bromo-1-thiazol-2-yl ethanone, 2-bromo-1-1,3-thiazol-2-yl ethanone, 2-bromoacetyl-1,3-thiazole, 2-bromo-1-thiazol-2-yl-ethanone, 2-bromo-1-1,3-thiazol-2-yl ethan-1-one, ethanone, 2-bromo-1-2-thiazolyl, 2-bromo-1-1,3-thiazol-2-yl-1-ethanone, 1-bromo-1,3-thiazole-2-yl ethane-2-one, 2-bromoacetylthiazole, 2-bromoacetyl thiazole
PubChem CID 2795212
IUPAC Name 2-bromo-1-(1,3-thiazol-2-yl)ethanone
SMILES BrCC(=O)C1=NC=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 206.06
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(1, 3-thiazol-2-yl)ethanone
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage 4°C