Learn More
2-Benzyloxyphenol, 98%
CAS: 6272-38-4 | C13H12O2 | 200.237 g/mol
Supplier: Thermo Scientific Chemicals A1902014
Description
2-Benzyloxyphenol is used in the synthesis of 2-(benzyloxy)hydroquinone and also used to prepare sequential polypeptides.It is used as reagent for synthesis of multidentate chelating ligands. It is also used in chemicals, pharmaceutical research and also acts a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Benzyloxyphenol is used in the synthesis of 2-(benzyloxy)hydroquinone and also used to prepare sequential polypeptides.It is used as reagent for synthesis of multidentate chelating ligands. It is also used in chemicals, pharmaceutical research and also acts a pharmaceutical intermediate.
Solubility
Soluble in alcohol, benzene, and diethyl ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
| 2-Benzyloxyphenol | |
| 1.143 | |
| C13H12O2 | |
| MFCD00002186 | |
| 2-benzyloxy phenol, 2-benzyloxyphenol, phenol, 2-phenylmethoxy, benzyloxyphenol, benzyl o-hydroxyphenyl ether, 2-phenylmethoxy phenol, 2-benzyloxy-phenol, o-benzyloxy phenol | |
| CCZCXFHJMKINPE-UHFFFAOYSA-N | |
| 2-phenylmethoxyphenol | |
| 80459 | |
| 98% |
| 6272-38-4 | |
| >110°C (230°F) | |
| 1.591 | |
| 25 g | |
| Soluble in alcohol,benzene,and diethyl ether. Insoluble in water. | |
| C1=CC=C(C=C1)COC2=CC=CC=C2O | |
| 200.237 | |
| 200.24 |
Chemical Identifiers
| 6272-38-4 | |
| 200.237 | |
| CCZCXFHJMKINPE-UHFFFAOYSA-N | |
| 80459 | |
| C1=CC=C(C=C1)COC2=CC=CC=C2O |
| C13H12O2 | |
| MFCD00002186 | |
| 2-benzyloxy phenol, 2-benzyloxyphenol, phenol, 2-phenylmethoxy, benzyloxyphenol, benzyl o-hydroxyphenyl ether, 2-phenylmethoxy phenol, 2-benzyloxy-phenol, o-benzyloxy phenol | |
| 2-phenylmethoxyphenol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 228-461-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only