accessibility menu, dialog, popup

UserName

CAS RN 6272-38-4

IUPAC Name: 2-(benzyloxy)phenol
Synonyms: 2-(benzyloxy)phenol
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: CCZCXFHJMKINPE-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)