missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxyphenol, 98%

Catalog No. p-7024411
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1902006 5 g
AAA1902014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1902006 Supplier Thermo Scientific Chemicals Supplier No. A1902006
Only null left

CAS: 6272-38-4 | C13H12O2 | 200.237 g/mol

2-Benzyloxyphenol is used in the synthesis of 2-(benzyloxy)hydroquinone and also used to prepare sequential polypeptides.It is used as reagent for synthesis of multidentate chelating ligands. It is also used in chemicals, pharmaceutical research and also acts a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzyloxyphenol is used in the synthesis of 2-(benzyloxy)hydroquinone and also used to prepare sequential polypeptides.It is used as reagent for synthesis of multidentate chelating ligands. It is also used in chemicals, pharmaceutical research and also acts a pharmaceutical intermediate.

Solubility
Soluble in alcohol, benzene, and diethyl ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 6272-38-4
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00002186
InChI Key CCZCXFHJMKINPE-UHFFFAOYSA-N
Synonym 2-benzyloxy phenol, 2-benzyloxyphenol, phenol, 2-phenylmethoxy, benzyloxyphenol, benzyl o-hydroxyphenyl ether, 2-phenylmethoxy phenol, 2-benzyloxy-phenol, o-benzyloxy phenol
PubChem CID 80459
IUPAC Name 2-phenylmethoxyphenol
SMILES C1=CC=C(C=C1)COC2=CC=CC=C2O

Specifications

Density 1.143
Flash Point >110°C (230°F)
Refractive Index 1.591
Quantity 5 g
Solubility Information Soluble in alcohol,benzene,and diethyl ether. Insoluble in water.
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material 2-Benzyloxyphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-461-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only