missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxybenzyl Acetate 98.0+%, TCI America™

Catalog No. B29425G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B29425G Supplier TCI America Supplier No. B29425G
Only null left

Chemical Identifiers

CAS 1073234-31-7
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
InChI Key AEQBBYFCUOIBIW-UHFFFAOYSA-N
Synonym Acetic Acid 2-Benzyloxybenzyl Ester
PubChem CID 44629830
IUPAC Name (2-phenylmethoxyphenyl)methyl acetate
SMILES CC(=O)OCC1=CC=CC=C1OCC2=CC=CC=C2

Specifications

Color Yellow
Quantity 5 g
Formula Weight 256.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Benzyloxybenzyl Acetate