missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxybenzaldehyde 98.0+%, TCI America™

Catalog No. B16535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16535G 5 g
B165325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16535G Supplier TCI America Supplier No. B16535G

Chemical Identifiers

CAS 5896-17-3
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00016583
InChI Key PBEJTRAJWCNHRS-UHFFFAOYSA-N
Synonym 2-benzyloxy benzaldehyde, 2-benzyloxybenzaldehyde, benzaldehyde, 2-phenylmethoxy, o-benzyloxy benzaldehyde, benzaldehyde, o-benzyloxy, o-benzylsalicylaldehyde, 2-benzyl-oxy benzaldehyde, 2-phenylmethoxy benzaldehyde, benzyloxybenzaldehyde, o-benzyloxybenzaldehyde
PubChem CID 344784
IUPAC Name 2-(benzyloxy)benzaldehyde
SMILES O=CC1=CC=CC=C1OCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 208°C
Quantity 5 g
Formula Weight 212.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Benzyloxybenzaldehyde