missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxy-4-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B37791G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B37791G Supplier TCI America Supplier No. B37791G
Only null left

Chemical Identifiers

CAS 848779-87-3
Molecular Formula C13H12BFO3
MDL Number MFCD03788402
InChI Key IFPQNCKVOQKEIU-UHFFFAOYSA-N
PubChem CID 2782665

Specifications

Melting Point 116°C
Color White
Quantity 1 g
Synonym 2-benzyloxy-4-fluorophenylboronic acid, 2-benzyloxy-4-fluorophenyl boronic acid, 2-benzyloxy-4-fluorobenzeneboronic acid, 4-fluoro-2-phenylmethoxyphenyl boronic acid, boronic acid, 4-fluoro-2-phenylmethoxy phenyl, boronic acid,b-4-fluoro-2-phenylmethoxy phenyl, pubchem5321
SMILES B(C1=C(C=C(C=C1)F)OCC2=CC=CC=C2)(O)O
IUPAC Name (4-fluoro-2-phenylmethoxyphenyl)boronic acid
Molecular Weight (g/mol) 246.044
Formula Weight 246.04
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzyloxy-4-fluorophenylboronic Acid (contains varying amounts of Anhydride)