missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyl-1,3-dioxolane 98.0+%, TCI America™

Catalog No. B140425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B140425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B140425ML Supplier TCI America Supplier No. B140425ML
Only null left

Chemical Identifiers

CAS 101-49-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00003215
InChI Key SSZACLYPEFCREM-UHFFFAOYSA-N
Synonym 2-benzyldioxolan, 1,3-dioxolane, 2-phenylmethyl, phenylacetaldehyde glycol acetal, phenylacetaldehyde ethylene acetal, 1,3-dioxolane, 2-benzyl, phenylacetaldehyde ethyleneglycol acetal, phenylacetaldehyde ethylene glycol acetal, 1,3-dioxolane,2-phenylmethyl, benzyldioxolan, 1, 2-benzyl
PubChem CID 7562
IUPAC Name 2-benzyl-1,3-dioxolane
SMILES C1COC(O1)CC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 mL
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Benzyl-1,3-dioxolane
RTECSNumber JH6768000
TSCA Yes