missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoylnaphthalene, 98%

CAS: 644-13-3 | C17H12O | 232.28 g/mol

$90.19 - $344.23

Chemical Identifiers

CAS 644-13-3
Molecular Formula C17H12O
Molecular Weight (g/mol) 232.28
MDL Number MFCD00004106
InChI Key SJNXJRVDSTZUFB-UHFFFAOYSA-N
Synonym 2-benzoylnaphthalene, 2-naphthyl phenyl ketone, 2-benzonaphthone, naphthalen-2-yl phenyl methanone, methanone, 2-naphthalenylphenyl, 2'-benzonaphthone, ketone, 2-naphthyl phenyl, 2-naphthylphenylketone, beta-benzoylnaphthalene, 2-naphthyl phenyl methanone
PubChem CID 69516
IUPAC Name naphthalen-2-yl(phenyl)methanone
SMILES O=C(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1812103
View Documents
Thermo Scientific Chemicals
A1812103
1 g
Each for $90.19
Only null left
 
AAA1812106
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
A1812106
5 g
Each for $344.23
Only null left
 
Description

Description

2-Benzoylnaphthalene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzoylnaphthalene is used as pharmaceutical intermediate.

Solubility
Soluble in hot methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

644-13-3
232.28
SJNXJRVDSTZUFB-UHFFFAOYSA-N
69516
O=C(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1
C17H12O
MFCD00004106
2-benzoylnaphthalene, 2-naphthyl phenyl ketone, 2-benzonaphthone, naphthalen-2-yl phenyl methanone, methanone, 2-naphthalenylphenyl, 2'-benzonaphthone, ketone, 2-naphthyl phenyl, 2-naphthylphenylketone, beta-benzoylnaphthalene, 2-naphthyl phenyl methanone
naphthalen-2-yl(phenyl)methanone

Specifications

644-13-3
216°C to 216°C (7 mmHg)
MFCD00004106
2-benzoylnaphthalene, 2-naphthyl phenyl ketone, 2-benzonaphthone, naphthalen-2-yl phenyl methanone, methanone, 2-naphthalenylphenyl, 2'-benzonaphthone, ketone, 2-naphthyl phenyl, 2-naphthylphenylketone, beta-benzoylnaphthalene, 2-naphthyl phenyl methanone
SJNXJRVDSTZUFB-UHFFFAOYSA-N
naphthalen-2-yl(phenyl)methanone
69516
98%
80°C to 82°C
C17H12O
1 g
Soluble in hot methanol.
O=C(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1
232.28
232.28
2-Benzoylnaphthalene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 211-410-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only