accessibility menu, dialog, popup

UserName

CAS RN 644-13-3

IUPAC Name: (naphthalen-2-yl)(phenyl)methanone
Synonyms: (naphthalen-2-yl)(phenyl)methanone
Molecular Weight (g/mol): 232.28
Molecular Formula: C17H12O
InChi Key: SJNXJRVDSTZUFB-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)