missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoylnaphthalene, 98%

CAS: 644-13-3 | C17H12O | 232.28 g/mol

Supplier:  Thermo Scientific Chemicals A1812103

 View more versions of this product

Catalog No. AAA1812103

Only null left
Explore available promotions

Description

Description

2-Benzoylnaphthalene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzoylnaphthalene is used as pharmaceutical intermediate.

Solubility
Soluble in hot methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-Benzoylnaphthalene
80°C to 82°C
C17H12O
1 g
Soluble in hot methanol.
O=C(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1
232.28
232.28
644-13-3
216°C to 216°C (7 mmHg)
MFCD00004106
2-benzoylnaphthalene, 2-naphthyl phenyl ketone, 2-benzonaphthone, naphthalen-2-yl phenyl methanone, methanone, 2-naphthalenylphenyl, 2'-benzonaphthone, ketone, 2-naphthyl phenyl, 2-naphthylphenylketone, beta-benzoylnaphthalene, 2-naphthyl phenyl methanone
SJNXJRVDSTZUFB-UHFFFAOYSA-N
naphthalen-2-yl(phenyl)methanone
69516
98%

Chemical Identifiers

644-13-3
232.28
SJNXJRVDSTZUFB-UHFFFAOYSA-N
69516
O=C(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1
C17H12O
MFCD00004106
2-benzoylnaphthalene, 2-naphthyl phenyl ketone, 2-benzonaphthone, naphthalen-2-yl phenyl methanone, methanone, 2-naphthalenylphenyl, 2'-benzonaphthone, ketone, 2-naphthyl phenyl, 2-naphthylphenylketone, beta-benzoylnaphthalene, 2-naphthyl phenyl methanone
naphthalen-2-yl(phenyl)methanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 211-410-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only