missing translation for 'onlineSavingsMsg'
Learn More

(-)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, > 95% ee, Thermo Scientific Chemicals

Catalog No. AC297040050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC297040050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC297040050 Supplier Thermo Scientific Chemicals Supplier No. 297040050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79200-56-9
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00211274
InChI Key DDUFYKNOXPZZIW-UHNVWZDZSA-N
Synonym 1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one, --1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one, 1s,4r-3-azabicyclo 2.2.1 hept-5-en-2-one, 2-azabicyclo 2,2,1 hept-5-en-3-one, vince lactam, 2-azabicyclo 2.2.1 hept-5-en-3-one, 1r,4s, --2-azabicyclo 2.2.1 hept-5-en-3-one, --vince lactam, 1s-+-2-azabicyclo 2.2.1 hept-5-en-3-one
PubChem CID 2725037
IUPAC Name (1S,4R)-3-azabicyclo[2.2.1]hept-5-en-2-one
SMILES C1C2C=CC1NC2=O

Specifications

Absorbance (c=1, CH2Cl2) (1 cm cell), (c=1, CH2Cl2) (1 cm cell) at 0nm, 0.1 max. at 400nm
CAS Min % 97.5
CAS Max % 100.0
Melting Point 92°C to 100°C
Color White to Beige
Enantiomeric Excess 95% min. (HPLC)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition − 560.00 (25.00°C c=1,CH2Cl2)
Specific Rotation -560
Formula Weight 109.13
Percent Purity 98%,>95% ee
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (-)-2-Azabicyclo[2.2.1]hept-5-en-3-one, >95%ee
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
May cause an allergic skin reaction.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

RUO – Research Use Only