missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiophene-3-carbonitrile, 97%, Thermo Scientific™

Catalog No. AAH6069903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6069903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6069903 Supplier Thermo Scientific Chemicals Supplier No. H6069903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 4651-82-5
Molecular Formula C5H4N2S
Molecular Weight (g/mol) 124.16
MDL Number MFCD00706298
InChI Key XVGHZFWFGXDIOU-UHFFFAOYSA-N
Synonym 2-amino-3-cyanothiophene, 2-amino-3-thiophenecarbonitrile, 2-amino-thiophene-3-carbonitrile, 3-thiophenecarbonitrile,2-amino, 3-thiophenecarbonitrile, 2-amino, 2-aminothiophen-3-carbonitril, pubchem5222, aminothiophenecarbonitrile, 2-amino-3-thienyl cyanide, 2-azanylthiophene-3-carbonitrile
PubChem CID 736607
IUPAC Name 2-aminothiophene-3-carbonitrile
SMILES NC1=C(C=CS1)C#N

Specifications

Melting Point 104°C to 108°C
Quantity 1 g
Formula Weight 124.16
Percent Purity 97%
Chemical Name or Material 2-Aminothiophene-3-carbonitrile
Hazard Category H302-H315-H318-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures