missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiazole, 97%

Catalog No. AC153895000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153895000 500 g Plastic bottle
AC153891000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC153895000 Supplier Thermo Scientific Chemicals Supplier No. 153895000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96-50-4 | C3H4N2S | 100.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96-50-4
Molecular Formula C3H4N2S
Molecular Weight (g/mol) 100.14
MDL Number MFCD00005325
InChI Key RAIPHJJURHTUIC-UHFFFAOYSA-N
Synonym 2-aminothiazole, aminothiazole, 2-thiazolamine, thiazol-2-amine, abadol, 2-thiazolylamine, abadole, basedol, 2-thiazylamine, 4-thiazolin-2-onimine
PubChem CID 2155
ChEBI CHEBI:40782
IUPAC Name 1,3-thiazol-2-amine
SMILES NC1=NC=CS1

Specifications

Melting Point 86°C to 91°C
Color Brown
Boiling Point 117°C (20.0 Mbar)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Quantity 500 g
Beilstein 27,155
Fieser 04,19
Merck Index 15,475
Solubility Information Solubility in water: 100g/L (20°C). Other solubilities: soluble in most common organic solvents,freely soluble in hcl and 20% h2so4
Formula Weight 100.14
Percent Purity 97%
Physical Form Crystalline Powder, Grains, Lumps or Flakes
Chemical Name or Material 2-Aminothiazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 202-511-6

RUO – Research Use Only