missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiazole, 97%

Catalog No. AAA1202636
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
2500 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1202636 500 g
AAA1202618 50 g
AAA1202622 100 g
AAA1202630 250 g
AAA120260E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AAA1202636 Supplier Thermo Scientific Chemicals Supplier No. A1202636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96-50-4 | C3H4N2S | 100.14 g/mol

2-Aminothiazole is used in the synthesis of 2-aminothiazole-modified silica gel and also used in Ulmann coupling with 2-chlorobenzoic acids mediated by ultrasonic irradiation. It acts as a component of disperse dye, thyroid inhibitor in the treatment of hyperthyroidism, as the acid tartrate and as a therapeutic drug for prion diseases. It is also used to kill bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, alcohol and ether.

Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, nitric acid and sulfuric acid.

Chemical Identifiers

CAS 96-50-4
Molecular Formula C3H4N2S
Molecular Weight (g/mol) 100.14
MDL Number MFCD00005325
InChI Key RAIPHJJURHTUIC-UHFFFAOYSA-N
Synonym 2-aminothiazole, aminothiazole, 2-thiazolamine, thiazol-2-amine, abadol, 2-thiazolylamine, abadole, basedol, 2-thiazylamine, 4-thiazolin-2-onimine
PubChem CID 2155
ChEBI CHEBI:40782
IUPAC Name 1,3-thiazol-2-amine
SMILES NC1=NC=CS1

Specifications

Melting Point 88°C to 92°C
Boiling Point 117°C (15 mmHg)
Quantity 500 g
Beilstein 105738
Merck Index 14,479
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 100.14
Percent Purity 97%
Chemical Name or Material 2-Aminothiazole
Hazard Category H302-H319-H335
Hazard Statement GHS H Statement
H341-H302
Suspected of causing genetic defects.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
EINECSNumber 202-511-6
RTECSNumber XJ2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only