missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoquinoline 97.0+%, TCI America™

Catalog No. A04175G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A04175G 5 g
A04171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A04175G Supplier TCI America Supplier No. A04175G

Chemical Identifiers

CAS 580-22-3
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00038079
InChI Key GCMNJUJAKQGROZ-UHFFFAOYSA-N
Synonym 2-aminoquinoline, 2-quinolinamine, quinolinamine, quinoline, 2-amino, quinolin-2-ylamine, 2-amino-quinoline, fragment 19, ccris 1677, chembl61236, 2-quinolylamine
PubChem CID 11379
IUPAC Name quinolin-2-amine
SMILES C1=CC=C2C(=C1)C=CC(=N2)N

Specifications

Melting Point 131°C
Quantity 5 g
Formula Weight 144.18
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoquinoline
RTECSNumber VA9621800
TSCA Yes