missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyridine-4-methanol, 97%

Catalog No. AAL1984603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1984603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1984603 Supplier Thermo Scientific Chemicals Supplier No. L1984603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 105250-17-7 | C6H9N2O | 125.15 g/mol

2-Aminopyridine-4-methanol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminopyridine-4-methanol is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 105250-17-7
Molecular Formula C6H9N2O
Molecular Weight (g/mol) 125.15
MDL Number MFCD03791261
InChI Key ZRJJXXDQIQFZBW-UHFFFAOYSA-O
Synonym 2-aminopyridin-4-yl methanol, 2-amino-pyridin-4-yl-methanol, 2-aminopyridine-4-methanol, 2-amino-4-pyridinylmethanol, 4-pyridinemethanol, 2-amino, 2-aminopyridin-4-yl-methanol, 2-amino-4-pyridinemethanol, 2-amino-4-pyridinyl methanol, 2-amino-4-hydroxymethylpyridine, 2-amino-4-pyridyl methan-1-ol
PubChem CID 1515296
IUPAC Name (2-aminopyridin-4-yl)methanol
SMILES NC1=CC(CO)=CC=[NH+]1

Specifications

Melting Point 81°C to 85°C
Quantity 1 g
Beilstein 8760550
Sensitivity Air sensitive
Solubility Information Soluble in water.
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 2-Aminopyridine-4-methanol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only