missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyrazine 98.0+%, TCI America™

Catalog No. A09895G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09895G 5 g
A098925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09895G Supplier TCI America Supplier No. A09895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5049-61-6
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.105
MDL Number MFCD00006137
InChI Key XFTQRUTUGRCSGO-UHFFFAOYSA-N
Synonym 2-aminopyrazine, aminopyrazine, pyrazinamine, 2-pyrazinamine, pyrazin-2-ylamine, pyrazine-2-ylamine, pyrazine, 2-amino, amino pyrazine, 3-aminopyrazine, iminopyrazine
PubChem CID 78747
IUPAC Name pyrazin-2-amine
SMILES C1=CN=C(C=N1)N

Specifications

Melting Point 121°C
Quantity 5 g
Formula Weight 95.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminopyrazine