missing translation for 'onlineSavingsMsg'
Learn More

2-Aminophenyl Phenyl Sulfide 98.0+%, TCI America™

Catalog No. A185625G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A185625G 25 g
A1856100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A185625G Supplier TCI America Supplier No. A185625G

Chemical Identifiers

CAS 1134-94-7
Molecular Formula C12H11NS
Molecular Weight (g/mol) 201.287
InChI Key DGBISJKLNVVJGD-UHFFFAOYSA-N
Synonym 2-Aminodiphenyl Sulfide, 2-(Phenylthio)aniline
PubChem CID 365393
IUPAC Name 2-phenylsulfanylaniline
SMILES C1=CC=C(C=C1)SC2=CC=CC=C2N

Specifications

Melting Point 43°C
Boiling Point 258°C
Quantity 25 g
UN Number 3077
Formula Weight 201.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminophenyl Phenyl Sulfide
EINECSNumber (3)-4188
TSCA No