missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminophenol 98.0+%, TCI America™
Supplier: TCI America A033525G
Specifications
| 2-Aminophenol | |
| 175°C | |
| MFCD00007690 | |
| 2512 | |
| CDAWCLOXVUBKRW-UHFFFAOYSA-N | |
| 2-aminophenol | |
| 5801 | |
| 109.13 | |
| Crystalline Powder |
| 95-55-6 | |
| C6H7NO | |
| 25 g | |
| o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga | |
| C1=CC=C(C(=C1)N)O | |
| 109.128 | |
| CHEBI:18112 | |
| ≥98.0% (T) |
Chemical Identifiers
| 95-55-6 | |
| 109.128 | |
| CDAWCLOXVUBKRW-UHFFFAOYSA-N | |
| 5801 | |
| 2-aminophenol |
| C6H7NO | |
| MFCD00007690 | |
| o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga | |
| CHEBI:18112 | |
| C1=CC=C(C(=C1)N)O |
Safety and Handling
EINECSNumber : (3)-0675
RTECSNumber : SJ4950000
TSCA : Yes