missing translation for 'onlineSavingsMsg'
Learn More

2-Aminophenol 98.0+%, TCI America™

Catalog No. p-7116843
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A033525G 25 g
A0335100G 100 g
A0335500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A033525G Supplier TCI America Supplier No. A033525G

Chemical Identifiers

CAS 95-55-6
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00007690
InChI Key CDAWCLOXVUBKRW-UHFFFAOYSA-N
Synonym o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga
PubChem CID 5801
ChEBI CHEBI:18112
IUPAC Name 2-aminophenol
SMILES C1=CC=C(C(=C1)N)O

Specifications

Melting Point 175°C
Quantity 25 g
UN Number 2512
Formula Weight 109.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminophenol
EINECSNumber (3)-0675
RTECSNumber SJ4950000
TSCA Yes