missing translation for 'onlineSavingsMsg'
Learn More

[2-(Aminomethyl)phenyl]acetic Acid 97.0+%, TCI America™

Catalog No. A21995G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21995G Supplier TCI America Supplier No. A21995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40851-65-8
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD04038440
InChI Key VLOIVYPDUSVCLZ-UHFFFAOYSA-N
PubChem CID 1514158
IUPAC Name 2-[2-(aminomethyl)phenyl]acetic acid
SMILES NCC1=CC=CC=C1CC(O)=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 165.19
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material [2-(Aminomethyl)phenyl]acetic Acid