missing translation for 'onlineSavingsMsg'
Learn More

2-Aminomethyl-1H-imidazole dihydrochloride, 97%

Catalog No. AC454590010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454590010 1 g
AC454590050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454590010 Supplier Thermo Scientific Chemicals Supplier No. 454590010

CAS: 22600-77-7 | C4H9Cl2N3 | 170.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22600-77-7
Molecular Formula C4H9Cl2N3
Molecular Weight (g/mol) 170.04
MDL Number MFCD06738779
InChI Key KYUDBQDDNKPSIC-UHFFFAOYSA-N
Synonym 1h-imidazol-2-yl methanamine dihydrochloride, 2-aminomethylimidazole dihydrochloride, 2-aminomethyl-1h-imidazole dihydrochloride, 1h-imidazol-2-ylmethylamine dihydrochloride, 1h-imidazol-2-ylmethanamine dihydrochloride, 1h-imidazole-2-methanamine dihydrochloride, 1h-imidazole-2-methanamine, dihydrochloride, 1-1h-imidazol-2-yl methanamine dihydrochloride, 1h-imidazol-2-ylmethyl amine dihydrochloride, 2-aminomethylimidazole hcl
PubChem CID 12417863
SMILES Cl.Cl.NCC1=NC=CN1

Specifications

Melting Point 250.0°C to 251.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 to max. 102.5% min. (Argentometry)
Packaging Glass Bottle
Quantity 1 g
IUPAC Name 1-(1H-imidazol-2-yl)methanamine dihydrochloride
Formula Weight 170.04
Percent Purity 98%
Chemical Name or Material 2-Aminomethyl-1H-imidazole dihydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only