missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminoisonicotinic Acid 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 290°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 138.13 |
| Percent Purity | ≥97.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Aminoisonicotinic Acid |
Safety and Handling
| TSCA | No |