missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethanol, ACS, 99%, Spectrum™ Chemical

Catalog No. 18608191
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
200 L
Packaging:
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608191 500 mL Poly Drum
18608192 1 L Poly Drum
18608189 2.5 L Poly Drum
18608193 4 L Poly Drum
18608188 20 L Poly Drum
18608190 200 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18608191 Supplier Spectrum Chemical Mfg Cor Supplier No. ET100500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
IUPAC Name 2-aminoethan-1-ol
SMILES NCCO

Specifications

CAS Max % 1
Boiling Point 170.8°C / 339.4°F
Packaging Poly Drum
Quantity 500 mL
Percent Purity 0.99
Grade ACS
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material 2-Aminoethanol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.