missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethanethiol 95.0+%, TCI America™

Catalog No. A0648500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0648500G 500 g
A064825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0648500G Supplier TCI America Supplier No. A0648500G
Only null left

Chemical Identifiers

CAS 60-23-1
Molecular Formula C2H7NS
Molecular Weight (g/mol) 77.15
MDL Number MFCD00008196
InChI Key UFULAYFCSOUIOV-UHFFFAOYSA-N
Synonym cysteamine, mercaptamine, thioethanolamine, becaptan, mercamine, cysteinamine, beta-mercaptoethylamine, 2-mercaptoethylamine, cysteamin, lambraten
PubChem CID 6058
ChEBI CHEBI:17141
IUPAC Name 2-aminoethane-1-thiol
SMILES NCCS

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 500 g
Formula Weight 77.15
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoethanethiol
EINECSNumber (9)-1422
RTECSNumber KJ0175000
TSCA No
Recommended Storage Refrigerator