missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzyl alcohol, 98%, Thermo Scientific™

Numéro de catalogue. AC153731000
Modifier l'affichage
Click to view available options
Quantité:
10g
25g
100g
Emballage:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC153731000 100g Glass bottle
AC153730100 10g Glass bottle
AC153730250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC153731000 Fournisseur Thermo Scientific Chemicals Code fournisseur 153731000

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 5344-90-1
Formule moléculaire C7H9NO
Poids moléculaire (g/mol) 123.15
Numéro MDL MFCD00007749
Clé InChI VYFOAVADNIHPTR-UHFFFAOYSA-N
Synonyme 2-aminobenzyl alcohol, 2-aminophenyl methanol, 2-aminobenzylalcohol, o-aminobenzyl alcohol, 2-aminobenzenemethanol, benzenemethanol, 2-amino, o-aminobenzylic alcohol, o-hydroxymethyl aniline, benzyl alcohol, o-amino, 2-hydroxymethylaniline
PubChem CID 21439
Nom de l’IUPAC (2-aminophenyl)methanol
SOURIRES C1=CC=C(C(=C1)CO)N

Spécifications

Nom ou substance chimique 2-Aminobenzyl alcohol
CAS % minimum 97.5
CAS % maximal 100.0
Point d’ébullition 162.0°C (15.0 mmHg)
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 97.5% min. (GC)
Emballage Glass bottle
Indice de réfraction 1.4905
Formule linéaire H2NC6H4CH2OH
Quantité 100g
Beilstein 13, 615
Renseignements sur la solubilité Solubility in water: 30g/100g (20°C). Other solubilities: 40g/100g thf (14°C),0.64g/100g toluene (20°C),soluble in chlorinated solvents,soluble in ethanol,ether,very soluble in benzene
Poids de formule 123.15
Pourcentage de pureté 98%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-293-7

RUO – Research Use Only