missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzaldehyde, 97%

Catalog No. AC279660010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC279660010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC279660010 Supplier Thermo Scientific Chemicals Supplier No. 279660010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 529-23-7 | C7H7NO | 121.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 529-23-7
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.14
MDL Number MFCD00007709
InChI Key FXWFZIRWWNPPOV-UHFFFAOYSA-N
Synonym o-aminobenzaldehyde, benzaldehyde, 2-amino, 2-amino-benzaldehyde, aminobenzaldehyde, anthranilaldehyde, 2-formylaniline, anthranil aldehyde, o-amino benzaldehyde, benzaldehyde, amino, pubchem15692
PubChem CID 68255
IUPAC Name 2-aminobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 37°C
Color White to Yellow
Boiling Point 80°C to 85°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula NH2C6H4CHO
Quantity 1 g
Formula Weight 121.14
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 2-Aminobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
EINECSNumber 208-454-3

RUO – Research Use Only