missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-m-cresol 98.0+%, TCI America™

Catalog No. A18515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18515G 5 g
A185125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18515G Supplier TCI America Supplier No. A18515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2835-97-4
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00075082
InChI Key FEDLEBCVFZMHBP-UHFFFAOYSA-N
Synonym 2-amino-m-cresol, phenol, 2-amino-3-methyl, 2-amino-3-methyl-phenol, 2-hydroxy-6-methylaniline, amino-m-cresol, ccris 8319, 2-azanyl-3-methyl-phenol, 2-hydroxy-6-methyl-aniline, acmc-1cp72, dsstox_cid_29117
PubChem CID 3014138
IUPAC Name 2-amino-3-methylphenol
SMILES CC1=CC=CC(O)=C1N

Specifications

Melting Point 151°C
Quantity 5 g
Formula Weight 123.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-m-cresol
EINECSNumber (3)-0707
TSCA No