missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methoxypyridine, 97%

Catalog No. AAH6643614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6643614 25 g
AAH6643606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6643614 Supplier Thermo Scientific Chemicals Supplier No. H6643614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17920-35-3 | C6H8N2O | 124.143 g/mol

It's an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It′s an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 17920-35-3
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD04972542
InChI Key DEUALFRBMNMGDS-UHFFFAOYSA-N
Synonym 2-amino-6-methoxypyridine, 6-methoxy-pyridin-2-ylamine, 6-methoxy-2-pyridinamine, 6-methoxypyrid-2-ylamine, 6-methoxy-2-pyridylamine, 2-amino-6-methoxy pyridine, 2-pyridinamine, 6-methoxy, methoxypyridin-6-amine, acmc-209efc, 6-methoxy-2-aminopyridin
PubChem CID 580053
IUPAC Name 6-methoxypyridin-2-amine
SMILES COC1=CC=CC(=N1)N

Specifications

Color Brown
Quantity 25 g
UN Number UN2810
Beilstein 115179
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 124.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Amino-6-methoxypyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only