missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chlorobenzothiazole 98.0+%, TCI America™

Catalog No. A121625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A121625G 25 g
A1216500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A121625G Supplier TCI America Supplier No. A121625G

Chemical Identifiers

CAS 95-24-9
Molecular Formula C7H5ClN2S
Molecular Weight (g/mol) 184.641
MDL Number MFCD00053557
InChI Key VMNXKIDUTPOHPO-UHFFFAOYSA-N
Synonym 2-amino-6-chlorobenzothiazole, 6-chlorobenzothiazol-2-ylamine, 2-benzothiazolamine, 6-chloro, 6-chlorobenzo d thiazol-2-amine, 6-chloro-benzothiazol-2-ylamine, unii-2u337t5ufg, 6-chloro-2-benzothiazolamine, benzothiazole, 2-amino-6-chloro, 6-chlorobenzothiazole-2-ylamine, c7h4brcln2s
PubChem CID 7226
IUPAC Name 6-chloro-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1Cl)SC(=N2)N

Specifications

Melting Point 199°C
Color White-Yellow
Quantity 25 g
Formula Weight 184.64
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-chlorobenzothiazole
EINECSNumber (5)-0236
RTECSNumber DL1575000
TSCA Yes