missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chlorobenzothiazole, 99%

Catalog No. AAH2787714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2787714 25 g
AAH2787706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2787714 Supplier Thermo Scientific Chemicals Supplier No. H2787714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95-24-9 | C7H5ClN2S | 184.641 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-24-9
Molecular Formula C7H5ClN2S
Molecular Weight (g/mol) 184.641
MDL Number MFCD00053557
InChI Key VMNXKIDUTPOHPO-UHFFFAOYSA-N
Synonym 2-amino-6-chlorobenzothiazole, 6-chlorobenzothiazol-2-ylamine, 2-benzothiazolamine, 6-chloro, 6-chlorobenzo d thiazol-2-amine, 6-chloro-benzothiazol-2-ylamine, unii-2u337t5ufg, 6-chloro-2-benzothiazolamine, benzothiazole, 2-amino-6-chloro, 6-chlorobenzothiazole-2-ylamine, c7h4brcln2s
PubChem CID 7226
IUPAC Name 6-chloro-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1Cl)SC(=N2)N

Specifications

Melting Point 199°C to 201°C
Quantity 25 g
Formula Weight 184.64
Percent Purity 99%
Chemical Name or Material 2-Amino-6-chlorobenzothiazole
Hazard Category H302
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 202-402-3
RTECSNumber DL1575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only