missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-phenylpyridine 98.0+%, TCI America™

Catalog No. A16671G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16671G 1 g
A1667100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16671G Supplier TCI America Supplier No. A16671G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33421-40-8
Molecular Formula C11H10N2
Molecular Weight (g/mol) 170.22
MDL Number MFCD01692452
InChI Key OAPVIBHQRYFYSE-UHFFFAOYSA-N
Synonym 2-amino-5-phenylpyridine, 5-phenyl-2-pyridinamine, 2-pyridinamine, 5-phenyl, phe-p-1, 5-phenyl-pyridin-2-ylamine, 5-phenylpyridin-2-ylamine, 2-amino-5-phenyl pyridine, pyridine, 2-amino-5-phenyl, acmc-1adxr, 5-phenyl-2-pyridylamine
PubChem CID 105097
IUPAC Name 5-phenylpyridin-2-amine
SMILES NC1=CC=C(C=N1)C1=CC=CC=C1

Specifications

Melting Point 135°C
Color Yellow
Quantity 1 g
Formula Weight 170.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-phenylpyridine
RTECSNumber US2145000
TSCA No