missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-phenylpyridine, 97%

Catalog No. AAH26002MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26002MD 250 mg
AAH2600203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH26002MD Supplier Thermo Scientific Chemicals Supplier No. H26002MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33421-40-8 | C11H10N2 | 170.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33421-40-8
Molecular Formula C11H10N2
Molecular Weight (g/mol) 170.22
MDL Number MFCD01692452
InChI Key OAPVIBHQRYFYSE-UHFFFAOYSA-N
Synonym 2-amino-5-phenylpyridine, 5-phenyl-2-pyridinamine, 2-pyridinamine, 5-phenyl, phe-p-1, 5-phenyl-pyridin-2-ylamine, 5-phenylpyridin-2-ylamine, 2-amino-5-phenyl pyridine, pyridine, 2-amino-5-phenyl, acmc-1adxr, 5-phenyl-2-pyridylamine
PubChem CID 105097
IUPAC Name 5-phenylpyridin-2-amine
SMILES NC1=CC=C(C=N1)C1=CC=CC=C1

Specifications

Melting Point 133°C to 137°C
Quantity 250 mg
Formula Weight 170.21
Percent Purity 97%
Chemical Name or Material 2-Amino-5-phenylpyridine
Hazard Category H315-H319-H335-H351
Hazard Statement GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
RTECSNumber US2145000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only