missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrobenzophenone, 98%

Catalog No. AAB2377306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2377306 5 g
AAB2377314 25 g
AAB2377322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2377306 Supplier Thermo Scientific Chemicals Supplier No. B2377306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1775-95-7 | C13H10N2O3 | 242.23 g/mol

It is used in the determination of nitrazepam in serum by gas-liquid chromatography; application in bioavailability studies and in second-harmonic generation in aromatic organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the determination of nitrazepam in serum by gas-liquid chromatography; application in bioavailability studies and in second-harmonic generation in aromatic organic compounds.

Solubility
Soluble in chloroform, dichloromethane, dimethyl sulfoxide, ethyl acetate and methanol. Soluble in water 3 mg/L (20°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1775-95-7
Molecular Formula C13H10N2O3
Molecular Weight (g/mol) 242.23
MDL Number MFCD00007364
InChI Key PZPZDEIASIKHPY-UHFFFAOYSA-N
Synonym 2-amino-5-nitrobenzophenone, 2-amino-5-nitrophenyl phenyl methanone, methanone, 2-amino-5-nitrophenyl phenyl, 2-amino-5-nitrophenyl phenylmethanone, 2-amino-5-nitrophenyl-phenylmethanone, 2-benzoyl-4-nitroaniline, 5-nitro-2-aminobenzophenone, 2-amino-5-nitro-phenyl-phenyl-methanone, 2-amino-5-nitrophenyl phenyl ketone, pubchem3254
PubChem CID 15681
IUPAC Name 2-benzoyl-4-nitroaniline
SMILES NC1=CC=C(C=C1C(=O)C1=CC=CC=C1)[N+]([O-])=O

Specifications

Melting Point 166°C to 168°C
Quantity 5 g
Solubility Information Soluble in chloroform,dichloromethane,dimethyl sulfoxide,ethyl acetate and methanol. Soluble in water 3mg/L (20°C).
Formula Weight 242.23
Percent Purity 98%
Chemical Name or Material 2-Amino-5-nitrobenzophenone
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 217-207-9
RTECSNumber PC4935000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only