missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methoxypyrimidine, 97%

Catalog No. AAH66799MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66799MD 250 mg
AAH6679903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66799MD Supplier Thermo Scientific Chemicals Supplier No. H66799MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13418-77-4 | C5H7N3O | 125.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13418-77-4
Molecular Formula C5H7N3O
Molecular Weight (g/mol) 125.13
MDL Number MFCD00187864
InChI Key KAHHAPNRIQLSFT-UHFFFAOYSA-N
PubChem CID 2305244
IUPAC Name 5-methoxypyrimidin-2-amine
SMILES COC1=CN=C(N)N=C1

Specifications

Color White
Quantity 250 mg
Beilstein 607358
Sensitivity Air Sensitive
Synonym 2-amino-5-methoxypyrimidine, 2-pyrimidinamine, 5-methoxy, 5-methoxy-2-pyrimidinamine, 2-pyrimidinamine, 5-methoxy-9ci, 5-methoxy-pyrimidin-2-ylamine, pubchem20801, enamine_000447, 2-pyrimidinamine,5-methoxy, 5-methoxypyrimidin-2-ylamine, ksc519a2h
Formula Weight 125.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Amino-5-methoxypyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only