missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluorobenzotrifluoride 97.0+%, TCI America™

Catalog No. A11585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11585G 5 g
A115825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11585G Supplier TCI America Supplier No. A11585G

Chemical Identifiers

CAS 393-39-5
Molecular Formula C7H5F4N
Molecular Weight (g/mol) 179.12
MDL Number MFCD00007831
InChI Key LRCQLCWUUBSUOY-UHFFFAOYSA-N
Synonym 2-amino-5-fluorobenzotrifluoride, 4-fluoro-2-trifluoromethyl aniline, 4-fluoro-2-trifluoromethyl-phenylamine, benzenamine, 4-fluoro-2-trifluoromethyl, alpha,alpha,alpha,4-tetrafluoro-o-toluidine, alpha, 2-amino-5-fluoro benzotrifluoride, 4-fluoro-2-trifluoromethyl phenylamine, alpha,alpha,alpha-4-tetrafluoro-o-toluidine, benzenamine,4-fluoro-2-trifluoromethyl
PubChem CID 67853
IUPAC Name 4-fluoro-2-(trifluoromethyl)aniline
SMILES NC1=CC=C(F)C=C1C(F)(F)F

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 179.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Amino-5-fluorobenzotrifluoride