missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzotrifluoride 98.0+%, TCI America™

Catalog No. A1058500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1058500G 500 g
A105825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1058500G Supplier TCI America Supplier No. A1058500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 445-03-4
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.57
MDL Number MFCD00007841
InChI Key CVINWVPRKDIGLL-UHFFFAOYSA-N
Synonym 2-amino-5-chlorobenzotrifluoride, 4-chloro-2-trifluoromethyl aniline, benzenamine, 4-chloro-2-trifluoromethyl, 2-amino-5-chlorobenzo trifluoride, 5-chloro-2-aminobenzotrifluoride, 2-amino-5-chloro-trifluorotoluene, 2-trifluoromethyl-4-chloro-aniline, 2-trifluoromethyl-4-chloroaniline, 4-chloro-alpha,alpha,alpha-trifluoro-o-toluidine, 4-chloro-2-trifluoromethyl-phenylamine
PubChem CID 67961
IUPAC Name 4-chloro-2-(trifluoromethyl)aniline
SMILES NC1=CC=C(Cl)C=C1C(F)(F)F

Specifications

Boiling Point 77°C
Quantity 500 g
Formula Weight 195.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Amino-5-chlorobenzotrifluoride