Learn More
2-Amino-5-chlorobenzoic acid, 98%
CAS: 635-21-2 | C7H6ClNO2 | 171.58 g/mol
Supplier: Thermo Scientific Chemicals A1432914
Description
2-Amino-5-chlorobenzoic acid is used in the preparation of disease-modifying antirheumatic drugs (DMARDs). Also used to produce 6-chloro-3H-quinazolin-4-one at temperature of 180°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-5-chlorobenzoic acid is used in the preparation of disease-modifying antirheumatic drugs (DMARDs). Also used to produce 6-chloro-3H-quinazolin-4-one at temperature of 180°C.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
| 2-Amino-5-chlorobenzoic acid | |
| ∼208°C (decomposition) | |
| MFCD00007838 | |
| 639027 | |
| Soluble in water. | |
| C1=CC(=C(C=C1Cl)C(=O)O)N | |
| 171.58 | |
| 171.58 |
| 635-21-2 | |
| C7H6ClNO2 | |
| 25 g | |
| 5-chloroanthranilic acid, 5-chloro-2-aminobenzoic acid, benzoic acid, 2-amino-5-chloro, anthranilic acid, 5-chloro, 2-amino-5-chloro-benzoic acid, unii-z2y66jmi31, 2-carboxy-4-chloroaniline, chembl36767, 5-amino-3-carboxy-1-chlorobenzene, 2-amino-5-chlorobenzoicacid | |
| IFXKXCLVKQVVDI-UHFFFAOYSA-N | |
| 2-amino-5-chlorobenzoic acid | |
| 12476 | |
| 98% |
Chemical Identifiers
| 635-21-2 | |
| 171.58 | |
| IFXKXCLVKQVVDI-UHFFFAOYSA-N | |
| 12476 | |
| C1=CC(=C(C=C1Cl)C(=O)O)N |
| C7H6ClNO2 | |
| MFCD00007838 | |
| 5-chloroanthranilic acid, 5-chloro-2-aminobenzoic acid, benzoic acid, 2-amino-5-chloro, anthranilic acid, 5-chloro, 2-amino-5-chloro-benzoic acid, unii-z2y66jmi31, 2-carboxy-4-chloroaniline, chembl36767, 5-amino-3-carboxy-1-chlorobenzene, 2-amino-5-chlorobenzoicacid | |
| 2-amino-5-chlorobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-230-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only