CAS RN 635-21-2
IUPAC Name:
2-amino-5-chlorobenzoic acid
Synonyms:
2-amino-5-chlorobenzoic acid
Molecular Weight (g/mol):
171.58
Molecular Formula:
C7H6ClNO2
InChi Key:
IFXKXCLVKQVVDI-UHFFFAOYSA-N
SMILES:
NC1=CC=C(Cl)C=C1C(O)=O
1
–
1
of
1
results
1
Promotions Available
2-Amino-5-chlorobenzoic acid, 98%
2-Amino-5-chlorobenzoic acid, 98%
CAS: 635-21-2 Molecular Formula: C7H6ClNO2 Molecular Weight (g/mol): 171.58 MDL Number: MFCD00007838 InChI Key: IFXKXCLVKQVVDI-UHFFFAOYSA-N Synonym: 5-chloroanthranilic acid,5-chloro-2-aminobenzoic acid,benzoic acid, 2-amino-5-chloro,anthranilic acid, 5-chloro,2-amino-5-chloro-benzoic acid,unii-z2y66jmi31,2-carboxy-4-chloroaniline,chembl36767,5-amino-3-carboxy-1-chlorobenzene,2-amino-5-chlorobenzoicacid PubChem CID: 12476 IUPAC Name: 2-amino-5-chlorobenzoic acid SMILES: C1=CC(=C(C=C1Cl)C(=O)O)N