missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzoic acid, 98%

Catalog No. p-7021832
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1432906 5 g
AAA1432914 25 g
AAA1432922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1432906 Supplier Thermo Scientific Chemicals Supplier No. A1432906

CAS: 635-21-2 | C7H6ClNO2 | 171.58 g/mol

2-Amino-5-chlorobenzoic acid is used in the preparation of disease-modifying antirheumatic drugs (DMARDs). Also used to produce 6-chloro-3H-quinazolin-4-one at temperature of 180°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-chlorobenzoic acid is used in the preparation of disease-modifying antirheumatic drugs (DMARDs). Also used to produce 6-chloro-3H-quinazolin-4-one at temperature of 180°C.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 635-21-2
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007838
InChI Key IFXKXCLVKQVVDI-UHFFFAOYSA-N
Synonym 5-chloroanthranilic acid, 5-chloro-2-aminobenzoic acid, benzoic acid, 2-amino-5-chloro, anthranilic acid, 5-chloro, 2-amino-5-chloro-benzoic acid, unii-z2y66jmi31, 2-carboxy-4-chloroaniline, chembl36767, 5-amino-3-carboxy-1-chlorobenzene, 2-amino-5-chlorobenzoicacid
PubChem CID 12476
IUPAC Name 2-amino-5-chlorobenzoic acid
SMILES C1=CC(=C(C=C1Cl)C(=O)O)N

Specifications

Melting Point ∼208°C (decomposition)
Quantity 5 g
Beilstein 639027
Solubility Information Soluble in water.
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material 2-Amino-5-chlorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-230-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only