missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloro-2'-fluorobenzophenone 98.0+%, TCI America™

Catalog No. A145125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A145125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A145125G Supplier TCI America Supplier No. A145125G

Chemical Identifiers

CAS 784-38-3
Molecular Formula C13H9ClFNO
Molecular Weight (g/mol) 249.669
MDL Number MFCD00038381
InChI Key GTGMXPIQRQSORU-UHFFFAOYSA-N
Synonym 2-amino-5-chloro-2'-fluorobenzophenone, 2-amino-5-chlorophenyl 2-fluorophenyl methanone, 2-amino-2'-fluoro-5-chlorobenzophenone, methanone, 2-amino-5-chlorophenyl 2-fluorophenyl, 4-chloro-2-2-fluorobenzoyl aniline, 2-amino-5-chloro-phenyl-2-fluorophenyl methanone, 2-amino-5-chlorophenyl 2-fluorophenyl ketone, 2-amino-5-chlorophenyl-2-fluorophenyl methanone, pubchem3252, acmc-1bd5r
PubChem CID 69912
IUPAC Name (2-amino-5-chlorophenyl)-(2-fluorophenyl)methanone
SMILES C1=CC=C(C(=C1)C(=O)C2=C(C=CC(=C2)Cl)N)F

Specifications

Melting Point 97°C
Color Yellow
Quantity 25 g
Formula Weight 249.67
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-chloro-2′-fluorobenzophenone