missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromopyrimidine, 97%

Catalog No. AAH2729106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2729106 5 g
AAH2729103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2729106 Supplier Thermo Scientific Chemicals Supplier No. H2729106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7752-82-1 | C4H4BrN3 | 174.001 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7752-82-1
Molecular Formula C4H4BrN3
Molecular Weight (g/mol) 174.001
MDL Number MFCD00012341
InChI Key UHRHPPKWXSNZLR-UHFFFAOYSA-N
Synonym 2-amino-5-bromopyrimidine, 5-bromopyrimidin-2-ylamine, 2-pyrimidinamine, 5-bromo, 5-bromo-2-aminopyrimidine, 5-bromo-2-pyrimidinamine, pyrimidine, 2-amino-5-bromo, 5-bromopyrimidine-2-amine, 2-amino-5-bromo-pyrimidine, 5-bromopyrimidine-2-ylamine, 5-bromo-pyrimidin-2-ylamine
PubChem CID 231310
IUPAC Name 5-bromopyrimidin-2-amine
SMILES C1=C(C=NC(=N1)N)Br

Specifications

Melting Point 232°C to 234°C
Quantity 5 g
Formula Weight 174
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromopyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only