missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-[(5-nitro-2-thiazolyl)thio]-1,3,4-thiadiazole 96.0+%, TCI America™
Supplier: TCI America A2940100MG
Specifications
| 2-Amino-5-[(5-nitro-2-thiazolyl)thio]-1,3,4-thiadiazole | |
| 160°C | |
| MFCD01927019 | |
| SU 3327 | |
| C1=C(SC(=N1)SC2=NN=C(S2)N)[N+](=O)[O-] | |
| 261.292 | |
| 261.29 | |
| Crystalline Powder |
| 40045-50-9 | |
| C5H3N5O2S3 | |
| 100 mg | |
| NQQBNZBOOHHVQP-UHFFFAOYSA-N | |
| 5-[(5-nitro-1,3-thiazol-2-yl)sulfanyl]-1,3,4-thiadiazol-2-amine | |
| 11837140 | |
| ≥96.0% (HPLC) |
Chemical Identifiers
| 40045-50-9 | |
| 261.292 | |
| NQQBNZBOOHHVQP-UHFFFAOYSA-N | |
| 11837140 | |
| C1=C(SC(=N1)SC2=NN=C(S2)N)[N+](=O)[O-] |
| C5H3N5O2S3 | |
| MFCD01927019 | |
| SU 3327 | |
| 5-[(5-nitro-1,3-thiazol-2-yl)sulfanyl]-1,3,4-thiadiazol-2-amine |
Safety and Handling
TSCA : No
Recommended Storage : Freezer