missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-[(5-nitro-2-thiazolyl)thio]-1,3,4-thiadiazole 96.0+%, TCI America™

Catalog No. p-7118157
Change view
Click to view available options
Quantity:
100 mg
20 mg
Catalog No. Quantity
A294020MG 20 mg
A2940100MG 100 mg
2 options

Catalog No. A294020MG

Supplier: TCI America A294020MG

Only null left

Chemical Identifiers

CAS 40045-50-9
Molecular Formula C5H3N5O2S3
Molecular Weight (g/mol) 261.292
MDL Number MFCD01927019
InChI Key NQQBNZBOOHHVQP-UHFFFAOYSA-N
Synonym SU 3327
PubChem CID 11837140
IUPAC Name 5-[(5-nitro-1,3-thiazol-2-yl)sulfanyl]-1,3,4-thiadiazol-2-amine
SMILES C1=C(SC(=N1)SC2=NN=C(S2)N)[N+](=O)[O-]

Specifications

Melting Point 160°C
Quantity 20 mg
Formula Weight 261.29
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-[(5-nitro-2-thiazolyl)thio]-1,3,4-thiadiazole
TSCA No
Recommended Storage Freezer